Please login to your online account to display your discounted pricing

(S)-(-)-α-Allylalanine hydrate, 99%, 99+% e.e., ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  275422500

 View more versions of this product

Catalog No. AC275422500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 129.16
Physical Form Needle-Like Crystalline Powder
Quantity 250mg
Color White
TSCA TSCA
Beilstein 04, 2852
CAS Max % 100.0
Packaging Glass bottle
Enantiomeric Excess (TLC):> 99.0%
Infrared Spectrum Authentic
Specific Rotation -17° to -19° (25°C, 589nm) (c=1, 6N HCl)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00149412
Name Note 99%, (99+% e.e.)
Chemical Name or Material (S)-(-)-α-Allylalanine hydrate
Linear Formula H2C=CHCH2CH(NH2)(CH3)CO2H·xH2O
CAS Min % 98.5
Assay Percent Range 99%
Molecular Formula C6 H11 N O2 . x H2 O
CAS 96886-55-4
Synonym L-α-Allylalanine
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (S)-(-)-α-Allylalanine hydrate
Name Note 99%, (99+% e.e.)
CAS 96886-55-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6 H11 N O2 . x H2 O
Linear Formula H2C=CHCH2CH(NH2)(CH3)CO2H·xH2O
Formula Weight 129.16
MDL Number MFCD00149412
Synonym L-α-Allylalanine
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents