Learn More
S-Acetylthiocholine iodide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102710050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
S-Acetylthiocholine iodide | |
1866-15-5 | |
White | |
97.5% min. (Argentometry) | |
C7H16INOS | |
MFCD00011819 | |
acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide | |
NTBLZMAMTZXLBP-UHFFFAOYSA-M | |
2-acetylsulfanylethyl(trimethyl)azanium;iodide | |
74629 | |
98% |
98% | |
205°C to 210°C | |
Authentic | |
Glass bottle | |
CH3COSCH2CH2N(CH3)3I | |
5 g | |
Solubility in water: freely soluble. Other solubilities: soluble in methanol | |
CC(=O)SCC[N+](C)(C)C.[I-] | |
289.17 | |
289.17 | |
Crystalline Powder |
Chemical Identifiers
1866-15-5 | |
NTBLZMAMTZXLBP-UHFFFAOYSA-M | |
74629 | |
CC(=O)SCC[N+](C)(C)C.[I-] |
MFCD00011819 | |
acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide | |
2-acetylsulfanylethyl(trimethyl)azanium;iodide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
GHS Signal Word: Danger
EINECSNumber : 217-474-1
RTECSNumber : FZ9865000
TSCA : TSCA
RUO – Research Use Only