Please login to your online account to display your discounted pricing

Alfa Aesar™ S-Acetamidomethyl-L-cysteine hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material S-Acetamidomethyl-L-cysteine hydrochloride
CAS 28798-28-9
Formula Weight 228.7
MDL Number MFCD00077080
Beilstein 4015821
Synonym H-Cys(Acm)-OH.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6317103

 
alfa aesar™
H6317103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6317106

 
alfa aesar™
H6317106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63171MD

 
alfa aesar™
H63171MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.7
Melting Point 165°C (dec.)
Chemical Name or Material S-Acetamidomethyl-L-cysteine hydrochloride
Physical Form Solid
Assay 98%
MDL Number MFCD00077080
CAS 28798-28-9
Beilstein 4015821
Synonym H-Cys(Acm)-OH.HCl
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C