missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Acetamidomethyl-L-cysteine hydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6317103
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
165°C (decomposition) | |
28798-28-9 | |
MFCD00077080 | |
h-cys acm-oh hcl, s-acetamidomethyl-l-cysteine hydrochloride, h-cys acm-oh.hcl, 2r-2-amino-3-acetamidomethyl sulfanyl propanoic acid hydrochloride, l-cysteine, s-acetylamino methyl-, monohydrochloride, h-cys acm .hcl, h-cys acm-oh hydrochloride, s-acetamidomethylcysteine hydrochloride, s-acetyl-aminomethyl-l-cysteine hydrochloride, h-cys acm-oh inverted exclamation mark currencyhcl | |
CC(=O)NCSCC(C(=O)O)N.Cl | |
228.691 | |
228.7 | |
1 g |
S-Acetamidomethyl-L-cysteine hydrochloride | |
C6H13ClN2O3S | |
4015821 | |
SZWPOAKLKGUXDD-JEDNCBNOSA-N | |
(2R)-3-(acetamidomethylsulfanyl)-2-aminopropanoic acid;hydrochloride | |
16211002 | |
98% | |
Solid |
Chemical Identifiers
28798-28-9 | |
228.691 | |
SZWPOAKLKGUXDD-JEDNCBNOSA-N | |
16211002 | |
CC(=O)NCSCC(C(=O)O)N.Cl |
C6H13ClN2O3S | |
MFCD00077080 | |
h-cys acm-oh hcl, s-acetamidomethyl-l-cysteine hydrochloride, h-cys acm-oh.hcl, 2r-2-amino-3-acetamidomethyl sulfanyl propanoic acid hydrochloride, l-cysteine, s-acetylamino methyl-, monohydrochloride, h-cys acm .hcl, h-cys acm-oh hydrochloride, s-acetamidomethylcysteine hydrochloride, s-acetyl-aminomethyl-l-cysteine hydrochloride, h-cys acm-oh inverted exclamation mark currencyhcl | |
(2R)-3-(acetamidomethylsulfanyl)-2-aminopropanoic acid;hydrochloride |
Safety and Handling
EINECSNumber : 249-230-5
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold