missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid, 97+%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54086MD

 View more versions of this product

Catalog No. AAH54086MD



Specifications

Specifications

(S)-(-)-9,10-Difluoro-3-methyl-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
>300°C
100986-89-8
MFCD04039905
NVKWWNNJFKZNJO-YFKPBYRVSA-N
281.215
281.22
−64° (c=1 in DMSO)
250mg
C13H9F2NO4
3s-9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O
688333
≥97%

Chemical Identifiers

100986-89-8
281.215
NVKWWNNJFKZNJO-YFKPBYRVSA-N
688333
C13H9F2NO4
MFCD04039905
3s-9,10-difluoro-2,3-dihydro-3-methyl-7-oxo-7h-pyrido 1,2,3-de-1,4-benzoxazine-6-carboxylic acid
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2F)F)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents