Please login to your online account to display your discounted pricing

Alfa Aesar™ S-[(6-Chloro-2-oxooxazolo[4,5-b]pyridin-3yl)methyl] O,O-dimethyl phosphorothioate, 95%

Chemical Identifiers

Chemical Name or Material S-[(6-Chloro-2-oxooxazolo[4,5-b]pyridin-3yl)methyl] O,O-dimethyl phosphorothioate
CAS 35575-96-3
Formula Weight 324.67
MDL Number MFCD00867611
Beilstein 1086470
Synonym Azamethiphos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6610503

  SDS
alfa aesar™
H6610503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6610506

  SDS
alfa aesar™
H6610506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66105MD

  SDS
alfa aesar™
H66105MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 324.67
Health Hazard 2 GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Physical Form Solid
Synonym Azamethiphos
CAS 35575-96-3
Health Hazard 1 Warning
Beilstein 1086470
Assay 95%
Storage Note 1 2° to 8°C
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
RTECS Number TE8070000
Chemical Name or Material S-[(6-Chloro-2-oxooxazolo[4,5-b]pyridin-3yl)methyl] O,O-dimethyl phosphorothioate
MDL Number MFCD00867611
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

2° to 8°C