missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole, TCI America™

Manufacturer:  TCI America P1784500MG

 View more versions of this product

Catalog No. P1784500MG



Specifications

Specifications

(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole
271°C
33878-70-5
XUHYQIQIENDJER-BYPYZUCNSA-N
5-[(2S)-pyrrolidin-2-yl]-2H-tetrazole
11275040
Crystal-Powder at 20°C
White-Yellow
500mg
C5H9N5
C1CC(NC1)C2=NNN=N2
139.162
139.16
≥98.0% (T)

Chemical Identifiers

33878-70-5
139.162
11275040
C1CC(NC1)C2=NNN=N2
C5H9N5
XUHYQIQIENDJER-BYPYZUCNSA-N
5-[(2S)-pyrrolidin-2-yl]-2H-tetrazole
Safety and Handling

Safety and Handling

TSCA : No

Documents