Please login to your online account to display your discounted pricing

(S)-(-)-5-Iodowillardiine, Tocris

Highly potent, subtype-selective kainate agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material (S)-(-)-5-Iodowillardiine
Assay Percent Range >98%
Molecular Formula C7H8IN3O4
Formula Weight 325.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

030710

 
r & d systems
030710
10mg Each for $239.63

030750

 
r & d systems
030750
50mg Each for $976.15
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 325.06
CAS 140187-25-3
Molecular Formula C7H8IN3O4
Assay Percent Range >98%
Chemical Name or Material (S)-(-)-5-Iodowillardiine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature