Please login to your online account to display your discounted pricing

(S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone p-toluenesulfonate, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC348410050

 View more versions of this product

Catalog No. AC348410050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 269.31
Quantity 5g
Color Colorless to beige
Melting Point 122° to 129°C
Specific Rotation Condition + 8.50 (20.00°C 589nm,(c=1,C2H5OH))
CAS Max % 100.0
CAS Min % 94.0
Specific Rotation +8° to +9° (25°C, 589nm) (c=1, C2H5OH)
Recommended Storage Refrigerator +4°C
Assay Percent Range 95%
Name Note 95%
MDL Number MFCD00274229
CAS 51693-17-5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone p-toluenesulfonate
Molecular Formula C12H15NO4S
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material (S)-(+)-5-(Hydroxymethyl)-2-pyrrolidinone p-toluenesulfonate
Name Note 95%
CAS 51693-17-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H15NO4S
Formula Weight 269.31
MDL Number MFCD00274229
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Refrigerator +4°C

SDS & Safety Documents