Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-5-Hydroxymethyl-2(5H)-furanone, 98%

$75.28 - $233.97

Chemical Identifiers

Chemical Name or Material (S)-(-)-5-Hydroxymethyl-2(5H)-furanone
CAS 78508-96-0
Molecular Formula C5H6O3
Formula Weight 114.1
MDL Number MFCD00075370
Beilstein 3537455
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL11919MD

 
alfa aesar™
L11919MD
250mg Each for $75.28

AAL1191903

 
alfa aesar™
L1191903
1g Each for $233.97
Description & Specifications

Specifications

Formula Weight 114.1
CAS 78508-96-0
Chemical Name or Material (S)-(-)-5-Hydroxymethyl-2(5H)-furanone
Assay 98%
Molecular Formula C5H6O3
MDL Number MFCD00075370
Beilstein 3537455
Melting Point 34° to 43°C
Quantity 250mg
Optical Rotation −148° (c=1.4 in Water)

Applications
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.