missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-5-Hydroxymethyl-2(5H)-furanone, 98%

Manufacturer:  Alfa Aesar™ L11919MD

 View more versions of this product

Catalog No. AAL11919MD



Description

Description

Applications
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone is used in the preparation of partially saturated heterocycles via a diastereoselective ring chain formation. It is also used in the synthesis of (1R,3S)-cis-chrysanthemic acid.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

(S)-(-)-5-Hydroxymethyl-2(5H)-furanone
34°C to 43°C
78508-96-0
MFCD00075370
s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, s---5-hydroxymethyl-2 5h-furanone, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one
C1=CC(=O)OC1CO
114.1
114.1
−148° (c=1.4 in Water)
250mg
C5H6O3
3537455
AWNLUIGMHSSXHB-BYPYZUCNSA-N
(2S)-2-(hydroxymethyl)-2H-furan-5-one
10176122
98%

Chemical Identifiers

78508-96-0
114.1
AWNLUIGMHSSXHB-BYPYZUCNSA-N
10176122
C1=CC(=O)OC1CO
C5H6O3
MFCD00075370
s---5-hydroxymethyl-2 5h-furanone, s-5-hydroxymethyl furan-2 5h-one, s---5-hydroxymethyl-2 5h-furanone, 2s-2-hydroxymethyl-2h-furan-5-one, 2 5h-furanone, 5-hydroxymethyl-, 5s, 5s-5-hydroxymethyl-5h-furan-2-one, pubchem7063, s-5-hydroxy-2-penten-4-olide, 5-hydroxymethyl-2 5h-furanone #, 5beta-hydroxymethyl furan-2 5h-one
(2S)-2-(hydroxymethyl)-2H-furan-5-one
Documents