Please login to your online account to display your discounted pricing

(S)-(-)-5-Fluorowillardiine, Tocris


Very potent AMPA agonist

Quantity 10mg

Manufacturer: r & d systems  030610

 View more versions of this product

Catalog No. 030610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 217.16
Recommended Storage Store at Room Temperature
Synonym (S)-(-)-α-Amino-5-fluoro-3, 4-dihydro-2, 4-dioxo-1(2H)pyridinepropanoic acid
Chemical Name or Material (S)-(-)-5-Fluorowillardiine
Molecular Formula C7H8FN3O4
Assay Percent Range >98%
CAS 140187-23-1

Specifications

Chemical Name or Material (S)-(-)-5-Fluorowillardiine
Assay Percent Range >98%
Molecular Formula C7H8FN3O4
Formula Weight 217.16
Synonym (S)-(-)-α-Amino-5-fluoro-3, 4-dihydro-2, 4-dioxo-1(2H)pyridinepropanoic acid
Accessories

Store at Room Temperature