missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-5-Fluorowillardiine, Tocris

Very potent AMPA agonist

$254.19 - $254.19

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C7H8FN3O4
Formula Weight 217.16
Synonym (S)-(-)-α-Amino-5-fluoro-3, 4-dihydro-2, 4-dioxo-1(2H)pyridinepropanoic acid
Chemical Name or Material (S)-(-)-5-Fluorowillardiine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
030610 R & D Systems
030610
10mg Each for $254.19
Specifications Description & Specifications

Chemical Identifiers

>98%
217.16
(S)-(-)-5-Fluorowillardiine
C7H8FN3O4
(S)-(-)-α-Amino-5-fluoro-3, 4-dihydro-2, 4-dioxo-1(2H)pyridinepropanoic acid

Specifications

10mg
(S)-(-)-α-Amino-5-fluoro-3, 4-dihydro-2, 4-dioxo-1(2H)pyridinepropanoic acid
>98%
(S)-(-)-5-Fluorowillardiine
217.16
C7H8FN3O4
140187-23-1

Safety and Handling

Recommended Storage : Store at Room Temperature