Please login to your online account to display your discounted pricing

(s)-(-)-[(5,6),(5',6')-bis(ethylenedioxy)biphenyl-2,2'-diyl](diphenylphosphine), 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-(-)-[(5,6),(5',6')-Bis(ethylenedioxy)biphenyl-2,2'-diyl](diphenylphosphine)
CAS 503538-68-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C40 H32 O4 P2
Formula Weight 638.64
MDL Number MFCD05861608
Synonym (S)-Synphos™
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375152500

  SDS
thermo fisher scientific
AC375152500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375151000

  SDS
thermo fisher scientific
AC375151000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 638.64
Quantity 250mg
Color White
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
CAS 503538-68-9
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C40 H32 O4 P2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Assay Percent Range 98%
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym (S)-Synphos™
MDL Number MFCD05861608
Chemical Name or Material (S)-(-)-[(5,6),(5',6')-Bis(ethylenedioxy)biphenyl-2,2'-diyl](diphenylphosphine)
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.