Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-4-tert-Butyloxazolidine-2,5-dione, 98%

$249.36 - $249.36

Chemical Identifiers

Chemical Name or Material (S)-(-)-4-tert-Butyloxazolidine-2,5-dione
CAS 62965-56-4
Molecular Formula C7H11NO3
Formula Weight 157.17
MDL Number MFCD05664067
Beilstein 4664497
Synonym L-tert-Leucine N-carboxyanhydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2022203

 
alfa aesar™
L2022203
1g Each for $249.36
Description & Specifications

Specifications

Formula Weight 157.17
Beilstein 4664497
Recommended Storage 2° to 8°C
Quantity 1g
Molecular Formula C7H11NO3
MDL Number MFCD05664067
CAS 62965-56-4
Chemical Name or Material (S)-(-)-4-tert-Butyloxazolidine-2,5-dione
Synonym L-tert-Leucine N-carboxyanhydride
Melting Point 126° to 128°C
Optical Rotation −41°
Assay 98%

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

2° to 8°C