Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-4-Phenyl-2-oxazolidinone, 98%

$119.61 - $370.97

Chemical Identifiers

Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone
CAS 99395-88-7
Molecular Formula C9H9NO2
Formula Weight 163.18
MDL Number MFCD00043396
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2730603

 
alfa aesar™
H2730603
1g Each for $119.61

AAH2730606

 
alfa aesar™
H2730606
5g Each for $370.97
Description & Specifications

Specifications

Formula Weight 163.18
MDL Number MFCD00043396
Quantity 1g
Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone
Assay 98%
Molecular Formula C9H9NO2
CAS 99395-88-7
Melting Point 130° to 132°C
Optical Rotation +52° (c=2 in Chloroform)