Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-4-Phenyl-2-butanol, 97%, ee 99%

$64.93 - $159.72

Chemical Identifiers

Chemical Name or Material (S)-(+)-4-Phenyl-2-butanol
CAS 22148-86-3
Molecular Formula C10H14O
Formula Weight 150.22
MDL Number MFCD03093082
Beilstein 4859487
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL18886MD

  SDS
alfa aesar™
L18886MD
250mg Each for $64.93

AAL1888603

  SDS
alfa aesar™
L1888603
1g Each for $159.72
Description & Specifications

Specifications

Formula Weight 150.22
Chemical Name or Material (S)-(+)-4-Phenyl-2-butanol
CAS 22148-86-3
Molecular Formula C10H14O
Assay 97%
Assay Percent Notes ee 99%
Beilstein 4859487
Boiling Point 239°C
Optical Rotation +14° (Neat)
Quantity 250mg
Flash Point >110°C (230°F)
MDL Number MFCD03093082