Please login to your online account to display your discounted pricing

(S)-(+)-4-Methyl-2-pentanol, 99%, ACROS Organics™

$257.35 - $257.35

Chemical Identifiers

Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
CAS 14898-80-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H14O
Formula Weight 102.18
MDL Number MFCD03093078
Synonym (S)-(+)-Methyl isobutyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC389230010

  SDS
thermo fisher scientific
AC389230010
Glass bottle 1g Each for $257.35
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 102.18
Quantity 1g
Color Clear colorless
Specific Rotation Condition +20° (20°C c=1,CHCl3)
Density 0.802g/mL
CAS Max % 100.0
Specific Rotation +17° to +23° (20°C, 589nm) (C=1, CHCl3)
Assay Percent Range 99%
Enantiomeric Excess 99.0% min.
MDL Number MFCD03093078
CAS 14898-80-7
Molecular Formula C6H14O
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
Refractive Index 1.4100 to 1.4120 (20°C, 589nm)
Packaging Glass bottle
Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
Infrared Spectrum Authentic
Synonym (S)-(+)-Methyl isobutyl carbinol
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.