missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ S-(4-Methoxybenzyl)-L-cysteine, tech. 80%

$60.46 - $272.96

Chemical Identifiers

CAS 2544-31-2
Molecular Formula C11H15NO3S
Molecular Weight (g/mol) 241.305
MDL Number MFCD00037207
InChI Key PQPZSPJVMUCVAQ-JTQLQIEISA-N
Synonym h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
PubChem CID 7009563
IUPAC Name (2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
SMILES COC1=CC=C(C=C1)CSCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6389406 View Documents Alfa Aesar™
H6389406
5g Each for $60.46
 
AAH6389414 View Documents Alfa Aesar™
H6389414
25g Each for $272.96
 
Specifications

Chemical Identifiers

2544-31-2
241.305
PQPZSPJVMUCVAQ-JTQLQIEISA-N
7009563
COC1=CC=C(C=C1)CSCC(C(=O)O)N
C11H15NO3S
MFCD00037207
h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
(2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid

Specifications

2544-31-2
MFCD00037207
PQPZSPJVMUCVAQ-JTQLQIEISA-N
(2R)-2-amino-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
7009563
Solid
Technical
S-(4-Methoxybenzyl)-L-cysteine
C11H15NO3S
h-cys pmeobzl-oh, h-cys 4-meo-bzl-oh, s-4-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxybenzyl thio propanoic acid, s-4-methoxybenzyl cysteine, s-p-methoxybenzyl-l-cysteine, r-2-amino-3-4-methoxy-benzylsulfanyl-propionic acid, 2-amino-3-4-methoxybenzyl thio propanoic acid, 2r-2-amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, ambotzhaa6100
COC1=CC=C(C=C1)CSCC(C(=O)O)N
241.305
241.31
80%
5g