Please login to your online account to display your discounted pricing

(S)-(-)-4-Hydroxy-2-pyrrolidinone, 98%, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398970010

 View more versions of this product

Catalog No. AC398970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
Quantity 1g
CAS 68108-18-9
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C4H7NO2
Formula Weight 101.1

Specifications

Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
Assay Percent Range 98%
Molecular Formula C4H7NO2
Formula Weight 101.1