Please login to your online account to display your discounted pricing

(S)-(-)-4-Hydroxy-2-pyrrolidinone, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
Assay Percent Range 98%
Molecular Formula C4H7NO2
Formula Weight 101.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC398970010

  SDS
thermo fisher scientific
AC398970010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC398970050

  SDS
thermo fisher scientific
AC398970050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
Quantity 1g
CAS 68108-18-9
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C4H7NO2
Formula Weight 101.1