Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(-)-4-Hydroxy-2-pyrrolidinone, ≥97%

Chemical Identifiers

Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
CAS 68108-18-9
Formula Weight 101.1
MDL Number MFCD00273370
Beilstein 1524192
Synonym (S)-β-Hydroxy-γ-butyrolactam
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66455MD

 
alfa aesar™
H66455MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6645503

 
alfa aesar™
H6645503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.1
Melting Point 156° to 159°C
MDL Number MFCD00273370
Assay ≥97%
Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone
CAS 68108-18-9
Physical Form Solid
Quantity 250mg
Synonym (S)-β-Hydroxy-γ-butyrolactam
Color White to off-white
Beilstein 1524192