missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5518306

 View more versions of this product

Catalog No. AAH5518306



Specifications

Specifications

(S)-4-Cyano-4'-(2-methylbutyl)biphenyl
63799-11-1
MFCD00192280
DNJQGRFZQMOYGM-AWEZNQCLSA-N
4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile
114240
97%
5g
C18H19N
1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl
CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
249.357
249.36

Chemical Identifiers

63799-11-1
249.357
DNJQGRFZQMOYGM-AWEZNQCLSA-N
114240
CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
C18H19N
MFCD00192280
1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl
4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents