missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Cyano-4'-(2-methylbutyl)biphenyl, 97%

Manufacturer:  Alfa Aesar™ H5518303

 View more versions of this product

Catalog No. AAH5518303



Specifications

Specifications

(S)-4-Cyano-4'-(2-methylbutyl)biphenyl
63799-11-1
MFCD00192280
DNJQGRFZQMOYGM-AWEZNQCLSA-N
4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile
114240
97%
1g
C18H19N
1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl
CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
249.357
249.36

Chemical Identifiers

63799-11-1
249.357
DNJQGRFZQMOYGM-AWEZNQCLSA-N
114240
CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
C18H19N
MFCD00192280
1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl
4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 264-464-8

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents