Please login to your online account to display your discounted pricing

Alfa Aesar™ S-(4-Chlorobenzyl)isothiouronium chloride, 98%

$183.82 - $516.82

Chemical Identifiers

Chemical Name or Material S-(4-Chlorobenzyl)isothiouronium chloride
CAS 544-47-8
Molecular Formula C8H10Cl2N2S
Formula Weight 237.15
MDL Number MFCD00012584
Merck Index 14,2128
Synonym 2-(4-Chlorobenzyl)-2-thiopseudourea hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2536214

  SDS
alfa aesar™
B2536214
25g Each for $183.82

AAB2536222

  SDS
alfa aesar™
B2536222
100g Each for $516.82
Description & Specifications

Specifications

Formula Weight 237.15
MDL Number MFCD00012584
Quantity 25g
Chemical Name or Material S-(4-Chlorobenzyl)isothiouronium chloride
TSCA TSCA
Assay 98%
CAS 544-47-8
Synonym 2-(4-Chlorobenzyl)-2-thiopseudourea hydrochloride
RTECS Number UM0950000
Molecular Formula C8H10Cl2N2S
Merck Index 14,2128
Melting Point 202° to 206°C