Please login to your online account to display your discounted pricing

(S)-4-Carboxyphenylglycine, Tocris

Competitive group I mGlu antagonist/weak group II agonist

$349.10 - $1,440.43

Chemical Identifiers

Chemical Name or Material (S)-4-Carboxyphenylglycine
Assay Percent Range >99%
Molecular Formula C9H9NO4
Formula Weight 195.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032310

 
r & d systems
RDS032310
10mg Each for $349.10

032350

 
r & d systems
RDS032350
50mg Each for $1,440.43
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 195.17
Chemical Name or Material (S)-4-Carboxyphenylglycine
Assay Percent Range >99%
Molecular Formula C9H9NO4
CAS 134052-73-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature