Please login to your online account to display your discounted pricing

(S)-4-Carboxy-3-hydroxyphenylglycine, Tocris


Group I antagonist/group II agonist

Quantity 50mg

Manufacturer: r & d systems  RDS032050

 View more versions of this product

Catalog No. 032050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 211.17
Chemical Name or Material (S)-4-Carboxy-3-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
CAS 85148-82-9
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material (S)-4-Carboxy-3-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.17
Accessories

Store at Room Temperature