Please login to your online account to display your discounted pricing

(S)-4-Carboxy-3-hydroxyphenylglycine, Tocris

Group I antagonist/group II agonist

$427.31 - $1,753.29

Chemical Identifiers

Chemical Name or Material (S)-4-Carboxy-3-hydroxyphenylglycine
Assay Percent Range >98%
Molecular Formula C9H9NO5
Formula Weight 211.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032010

 
r & d systems
RDS032010
10mg Each for $427.31

032050

 
r & d systems
RDS032050
50mg Each for $1,753.29
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 211.17
Molecular Formula C9H9NO5
Chemical Name or Material (S)-4-Carboxy-3-hydroxyphenylglycine
Assay Percent Range >98%
CAS 85148-82-9
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature