missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Boc-(3-hydroxymethyl)morpholine, 97%

$85.14 - $1,238.32

Chemical Identifiers

CAS 714971-28-5
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD06799478
InChI Key AIQSXVGBMCJQAG-QMMMGPOBSA-N
Synonym s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
PubChem CID 1514266
IUPAC Name tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CCOCC1CO
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64574MD View Documents Alfa Aesar™
H64574MD
250mg Each for $85.14
 
AAH6457403 View Documents Alfa Aesar™
H6457403
1g Each for $269.86
 
AAH6457406 View Documents Alfa Aesar™
H6457406
5g Each for $1,238.32
 
Description

Description

Applications
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

714971-28-5
217.265
AIQSXVGBMCJQAG-QMMMGPOBSA-N
1514266
CC(C)(C)OC(=O)N1CCOCC1CO
C10H19NO4
MFCD06799478
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate

Specifications

White
250mg
C10H19NO4
10251196
AIQSXVGBMCJQAG-QMMMGPOBSA-N
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
1514266
Powder
(S)-4-Boc-(3-hydroxymethyl)morpholine
80°C (literature)
714971-28-5
MFCD06799478
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate, s-4-boc-3-hydroxymethyl morpholine, s-n-boc-3-hydroxymethyl morpholine, tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate, s-n-boc-3-hydroxymethylmorpholine, 3 s-hydroxymethyl-4-bocmorpholine, s-4-boc-3-hydroxymethylmorpholine, s-4-boc-3-hydroxymethyl-morpholine, s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester, pubchem5634
CC(C)(C)OC(=O)N1CCOCC1CO
217.265
217.27
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C