missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals

$185.88 - $727.84

Chemical Identifiers

CAS 252049-08-4
Molecular Formula C27H28N2O5
Molecular Weight (g/mol) 460.53
MDL Number MFCD02259487
InChI Key KKUPBFMKUPMFRE-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
PubChem CID 70700347
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
SMILES C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6372603
View Documents Promotion Details
Thermo Scientific Chemicals
H6372603
1 g
Each for $185.88
 
AAH6372606
View Documents Promotion Details
Thermo Scientific Chemicals
H6372606
5 g
Each for $727.84
 
Description

Description

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

252049-08-4
460.53
KKUPBFMKUPMFRE-VWLOTQADSA-N
70700347
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C27H28N2O5
MFCD02259487
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid

Specifications

252049-08-4
C27H28N2O5
1 g
Slightly soluble in water.
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
460.53
474.51
Powder
White
MFCD02259487
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
KKUPBFMKUPMFRE-VWLOTQADSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
70700347
95%
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only