missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6372603
Description
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid | |
White | |
MFCD02259487 | |
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid | |
KKUPBFMKUPMFRE-VWLOTQADSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid | |
70700347 | |
95% |
252049-08-4 | |
C27H28N2O5 | |
1 g | |
Slightly soluble in water. | |
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
460.53 | |
474.51 | |
Powder |
Chemical Identifiers
252049-08-4 | |
460.53 | |
KKUPBFMKUPMFRE-VWLOTQADSA-N | |
70700347 | |
C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C27H28N2O5 | |
MFCD02259487 | |
s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only