Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Benzyl-3-butyryl-2-oxazolidinone

Chemical Identifiers

Chemical Name or Material (S)-4-Benzyl-3-butyryl-2-oxazolidinone
CAS 112459-79-7
Formula Weight 247.29
MDL Number MFCD06411406
Synonym UIC-1017; Cell Sheet Migration Inhibitor, Negative Control
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64844LB0

 
alfa aesar™
J64844LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.29
Chemical Name or Material (S)-4-Benzyl-3-butyryl-2-oxazolidinone
MDL Number MFCD06411406
CAS 112459-79-7
Physical Form Powder
Synonym UIC-1017; Cell Sheet Migration Inhibitor, Negative Control
Color White
Storage Note 1 2° to 8°C
Quantity 5mg

  • A cell-permeable N-(butyryl)oxazolidinone compound that serves as a negative control for the Cell Sheet Migration Inhibitor

Safety and Handling

2° to 8°C