Please login to your online account to display your discounted pricing

(S)-4-Benzyl-2-oxazolidinone, 99%, >99% ee, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC402161000

 View more versions of this product

Catalog No. AC402161000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material (S)-4-Benzyl-2-oxazolidinone
CAS 90719-32-7
Enantiomeric Excess 99% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Specific Rotation −60° to −64° (20°C, 589nm) (c=1, CHCl3)
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C10H11NO2
Specific Rotation Condition − 62.00 (20.00°C C=1,CHCl3)
Formula Weight 177.2
Melting Point 87° to 90°C
Color White to very slightly yellow
Quantity 100g

Specifications

Chemical Name or Material (S)-4-Benzyl-2-oxazolidinone
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C10H11NO2
Formula Weight 177.2