missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(-)-4-Amino-2-hydroxybutyric acid, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 342430250

 View more versions of this product

Catalog No. AC342430250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

(S)-(-)-4-Amino-2-hydroxybutyric acid
Glass bottle
White to Beige
25g
40371-51-5
100.0
C4H9NO3
MFCD00674110
IVUOMFWNDGNLBJ-VKHMYHEASA-N
(2S)-4-amino-2-hydroxybutanoic acid
2733929
Crystalline Powder, Crystals and/or Chunks
Authentic
-30
200°C to 203°C
96%
95.0
96%
H2N(CH2)2CH(OH)COOH
2s-4-amino-2-hydroxybutanoic acid, butanoic acid, 4-amino-2-hydroxy-, 2s, l-2-hydroxy-4-aminobutyric acid, pubchem23215, s---4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyricacid, s---amino-2-hydroxybutyric acid, s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid
C(CN)C(C(=O)O)O
119.12
119.12
96%

Chemical Identifiers

40371-51-5
119.12
IVUOMFWNDGNLBJ-VKHMYHEASA-N
2733929
C(CN)C(C(=O)O)O
C4H9NO3
MFCD00674110
2s-4-amino-2-hydroxybutanoic acid, butanoic acid, 4-amino-2-hydroxy-, 2s, l-2-hydroxy-4-aminobutyric acid, pubchem23215, s---4-amino-2-hydroxybutyric acid, s---4-amino-2-hydroxybutyricacid, s---amino-2-hydroxybutyric acid, s-4-amino-2-hydroxybutanoic acid, s-4-amino-2-hydroxybutyric acid
(2S)-4-amino-2-hydroxybutanoic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents