missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Amino-2-(Boc-amino)butyric acid, 97%

Manufacturer:  Alfa Aesar™ H62450MD

 View more versions of this product

Catalog No. AAH62450MD



Specifications

Specifications

(S)-4-Amino-2-(Boc-amino)butyric acid
250mg
C9H18N2O4
4745487
MDCPCLPRWLKUIQ-LURJTMIESA-N
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
7019666
Powder
White
25691-37-6
MFCD00236841
boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
CC(C)(C)OC(=O)NC(CCN)C(=O)O
218.253
218.25
97%

Chemical Identifiers

25691-37-6
218.253
MDCPCLPRWLKUIQ-LURJTMIESA-N
7019666
CC(C)(C)OC(=O)NC(CCN)C(=O)O
C9H18N2O4
MFCD00236841
boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P310-P362-P321-P332+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.

Danger

Documents