Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid, 95%

$53.19 - $53.19

Chemical Identifiers

Chemical Name or Material (S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid
CAS 62234-40-6
Formula Weight 252.27
MDL Number MFCD00077904
Beilstein 5960816
Synonym Z-Dab-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6579103

  SDS
alfa aesar™
H6579103
1g Each for $53.19

AAH6579106

  SDS
alfa aesar™
H6579106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 252.27
Melting Point 194° to 196°C
CAS 62234-40-6
Chemical Name or Material (S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid
Assay 95%
Beilstein 5960816
Physical Form Powder
Storage Note 1 2° to 8°C
Quantity 1g
Color Pale brown
MDL Number MFCD00077904
Synonym Z-Dab-OH
Safety and Handling

2° to 8°C