missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-(4-Aminobenzyl)-2-oxazolidinone, 97%

Manufacturer:  Alfa Aesar™ H5601703

 View more versions of this product

Catalog No. AAH5601703



Description

Description

Applications
(S)-4-(4-Aminobenzyl)-2-(1H)-oxazolidinone is an intermediate of zolmitriptan (sc-220415) which acts as a novel inhibitor of the cytochrome P-450 enzyme aromatase.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store at -20°C. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Specifications

(S)-4-(4-Aminobenzyl)-2-oxazolidinone
1g
C10H12N2O2
2-oxazolidinone, 4-4-aminophenyl methyl-, 4s
C1C(NC(=O)O1)CC2=CC=C(C=C2)N
192.218
192.22
107°C to 111°C
152305-23-2
MFCD03411476
WNAVSKJKDPLWBD-VIFPVBQESA-N
(4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
7099156
97%

Chemical Identifiers

152305-23-2
192.218
WNAVSKJKDPLWBD-VIFPVBQESA-N
7099156
C1C(NC(=O)O1)CC2=CC=C(C=C2)N
C10H12N2O2
MFCD03411476
2-oxazolidinone, 4-4-aminophenyl methyl-, 4s
(4S)-4-[(4-aminophenyl)methyl]-1,3-oxazolidin-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents