Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS

Asymmetric hydrogenation

$376.31 - $376.31

Chemical Identifiers

Chemical Name or Material (S)- (+)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane
CAS 325168-88-5
Name Note CTH-(S)-3,5-xylyl-PHANEPHOS
Molecular Formula C48H50P2
Formula Weight 688.87g/mol
MDL Number MFCD03840578
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4461602

 
alfa aesar™
44616-02
0.5g Each for $376.31
Description & Specifications

Specifications

Color White
Quantity 0.5g
Odor Odorless
Chemical Name or Material (S)- (+)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane
Name Note CTH-(S)-3,5-xylyl-PHANEPHOS
CAS 325168-88-5
Physical Form Powder
Molecular Formula C48H50P2
Formula Weight 688.87g/mol
MDL Number MFCD03840578
Storage Note 1
  • Packaged under Argon

  • Packaged under Argon

Safety and Handling

  • Packaged under Argon