missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-4-(1-Aminoethyl)phenol, 97%

Manufacturer:  Alfa Aesar™ H63269MD

 View more versions of this product

Catalog No. AAH63269MD



Description

Description

Applications
(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

(S)-4-(1-Aminoethyl)phenol
250mg
C8H11NO
UN3259
CDQPLIAKRDYOCB-LURJTMIESA-N
4-[(1S)-1-aminoethyl]phenol
41097924
Solid
White
221670-72-0
MFCD03844646
4-1s-1-aminoethyl phenol, s-4-1-aminoethyl phenol, phenol, 4-1s-1-aminoethyl, s-4-1-aminoethyl-phenol, s-4-1-amino-ethyl-phenol, s-4-1-amino-aethyl-phenol, phenol,4-1s-1-aminoethyl, s-4-hydroxy-alpha-methylbenzylamine, phenol, 4-1s-1-aminoethyl-9ci, s-4-hydroxy-? inverted exclamation marka-methylbenzylamine
CC(C1=CC=C(C=C1)O)N
137.182
137.18
97%

Chemical Identifiers

221670-72-0
137.182
CDQPLIAKRDYOCB-LURJTMIESA-N
41097924
CC(C1=CC=C(C=C1)O)N
C8H11NO
MFCD03844646
4-1s-1-aminoethyl phenol, s-4-1-aminoethyl phenol, phenol, 4-1s-1-aminoethyl, s-4-1-aminoethyl-phenol, s-4-1-amino-ethyl-phenol, s-4-1-amino-aethyl-phenol, phenol,4-1s-1-aminoethyl, s-4-hydroxy-alpha-methylbenzylamine, phenol, 4-1s-1-aminoethyl-9ci, s-4-hydroxy-? inverted exclamation marka-methylbenzylamine
4-[(1S)-1-aminoethyl]phenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

storageNote1 : 2° to 8°C

SDS Documents