Please login to your online account to display your discounted pricing

(S)-3-tert-Butyl-2,5-piperazinedione, 95%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  297845000

 View more versions of this product

Catalog No. AC297845000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 170.21
Physical Form Crystalline Powder
Quantity 500mg
Color Off-white to light brown
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 MOISTURE SENSITIVE
CAS 65050-07-9
Molecular Formula C8 H14 N2 O2
MDL Number MFCD00077675
Assay Percent Range 95%
Name Note 95%
Chemical Name or Material (S)-3-tert-Butyl-2, 5-piperazinedione
Packaging Glass bottle
Health Hazard 1 LIGHT SENSITIVE

Specifications

Chemical Name or Material (S)-3-tert-Butyl-2, 5-piperazinedione
Name Note 95%
CAS 65050-07-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8 H14 N2 O2
Formula Weight 170.21
MDL Number MFCD00077675
Accessories

LIGHT SENSITIVE

MOISTURE SENSITIVE

SDS & Safety Documents