Please login to your online account to display your discounted pricing

(S)-3-tert-Butyl-2,5-piperazinedione, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (S)-3-tert-Butyl-2, 5-piperazinedione
Name Note 95%
CAS 65050-07-9
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8 H14 N2 O2
Formula Weight 170.21
MDL Number MFCD00077675
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC297845000

  SDS
thermo fisher scientific
297845000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 170.21
Quantity 500mg
Color Off-white to light brown
TSCA TSCA
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material (S)-3-tert-Butyl-2, 5-piperazinedione
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C8 H14 N2 O2
MDL Number MFCD00077675
CAS 65050-07-9
Health Hazard 2 MOISTURE SENSITIVE
Packaging Glass bottle
Health Hazard 1 LIGHT SENSITIVE
Safety and Handling

LIGHT SENSITIVE

MOISTURE SENSITIVE