missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic Acid Hydrate, TCI America™

$155.84 - $534.73

Chemical Identifiers

CAS 162558-25-0
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00235896
InChI Key PKAUMAVONPSDRW-IBGZPJMESA-N
Synonym fmoc-dap boc-oh, n-fmoc-n'-boc-l-2,3-diaminopropionic acid, fmoc-dpr boc-oh, fmoc-l-2,3-diaminopropionic acid boc, fmoc-alpha,beta-diaminopropionic acid boc-oh, 2s-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-dap boc, fmoc-dap boc, fmoc-l-dpa boc-oh, ambotzfaa1367
PubChem CID 2756103
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B41741G View Documents TCI America
B41741G
1g Each for $155.84
 
B41745G View Documents TCI America
B41745G
5g Each for $534.73
 
Specifications

Chemical Identifiers

162558-25-0
426.469
PKAUMAVONPSDRW-IBGZPJMESA-N
2756103
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C23H26N2O6
MFCD00235896
fmoc-dap boc-oh, n-fmoc-n'-boc-l-2,3-diaminopropionic acid, fmoc-dpr boc-oh, fmoc-l-2,3-diaminopropionic acid boc, fmoc-alpha,beta-diaminopropionic acid boc-oh, 2s-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-dap boc, fmoc-dap boc, fmoc-l-dpa boc-oh, ambotzfaa1367
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

White
1g
C23H26N2O6
fmoc-dap boc-oh, n-fmoc-n'-boc-l-2,3-diaminopropionic acid, fmoc-dpr boc-oh, fmoc-l-2,3-diaminopropionic acid boc, fmoc-alpha,beta-diaminopropionic acid boc-oh, 2s-3-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-l-dap boc, fmoc-dap boc, fmoc-l-dpa boc-oh, ambotzfaa1367
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
426.469
426.47
≥97.0% (HPLC,T)
63°C
162558-25-0
MFCD00235896
PKAUMAVONPSDRW-IBGZPJMESA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
2756103
Crystal-Powder at 20°C
(S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic Acid Hydrate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator