Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-(+)-3-Octanol, 96%

Chemical Identifiers

Chemical Name or Material (S)-(+)-3-Octanol
CAS 22658-92-0
Formula Weight 130.23
MDL Number MFCD06797294
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6696306

 
alfa aesar™
H6696306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6696314

 
alfa aesar™
H6696314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.23
Boiling Point 174° to 177°C
Chemical Name or Material (S)-(+)-3-Octanol
CAS 22658-92-0
Physical Form Liquid
Assay 96%
MDL Number MFCD06797294
Density 0.819g/mL
Refractive Index 1.4250
Color Colorless to yellow
Quantity 5g