missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(+)-3-Octanol, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6696306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(S)-(+)-3-Octanol | |
Colorless to Yellow | |
174°C to 177°C | |
C8H18O | |
MFCD06797294 | |
s-+-3-octanol, 3s-octan-3-ol, octan-3s-ol, s-octan-3-ol, 3s-3-octanol, 3-octanol, 3s, unii-73dz0u3u1e component | |
CCCCCC(CC)O | |
130.231 | |
130.23 | |
Liquid |
22658-92-0 | |
0.819 g/mL | |
68°C (154°F) | |
1.425 | |
5 g | |
NMRPBPVERJPACX-QMMMGPOBSA-N | |
(3S)-octan-3-ol | |
6999002 | |
96% |
Chemical Identifiers
22658-92-0 | |
130.231 | |
NMRPBPVERJPACX-QMMMGPOBSA-N | |
6999002 | |
CCCCCC(CC)O |
C8H18O | |
MFCD06797294 | |
s-+-3-octanol, 3s-octan-3-ol, octan-3s-ol, s-octan-3-ol, 3s-3-octanol, 3-octanol, 3s, unii-73dz0u3u1e component | |
(3S)-octan-3-ol |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures