Please login to your online account to display your discounted pricing

Alfa Aesar™ (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid
CAS 339531-50-9
Formula Weight 480.56
MDL Number MFCD03490501
Synonym Fmoc-Leu-Ser(Psi(Me,Me)pro)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6662903

 
alfa aesar™
H6662903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66629MD

 
alfa aesar™
H66629MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 480.56
Chemical Name or Material (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid
Physical Form Solid
Assay 95%
MDL Number MFCD03490501
CAS 339531-50-9
Synonym Fmoc-Leu-Ser(Psi(Me,Me)pro)-OH
Quantity 1g