missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-3-(Methylamino)-1-(2-thienyl)-1-propanol 98.0+%, TCI America™

$128.38 - $383.48

Chemical Identifiers

CAS 116539-55-0
Molecular Formula C8H13NOS
Molecular Weight (g/mol) 171.258
MDL Number MFCD07357308
InChI Key YEJVVFOJMOHFRL-ZETCQYMHSA-N
Synonym (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene
PubChem CID 10095047
IUPAC Name (1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol
SMILES CNCCC(C1=CC=CS1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21575G View Documents Tci America
M21575G
5g Each for $128.38
 
M215725G View Documents Tci America
M215725G
25g Each for $383.48
 
Specifications

Chemical Identifiers

116539-55-0
171.258
YEJVVFOJMOHFRL-ZETCQYMHSA-N
10095047
CNCCC(C1=CC=CS1)O
C8H13NOS
MFCD07357308
(S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene
(1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol

Specifications

Undesignated
5g
C8H13NOS
(S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene
CNCCC(C1=CC=CS1)O
171.258
171.26
≥98.0% (GC,T)
74°C
116539-55-0
MFCD07357308
YEJVVFOJMOHFRL-ZETCQYMHSA-N
(1S)-3-(methylamino)-1-thiophen-2-ylpropan-1-ol
10095047
Crystal-Powder at 20°C
(S)-3-(Methylamino)-1-(2-thienyl)-1-propanol
Safety and Handling

Safety and Handling

TSCA : No