missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-3-Methyl-1-pentanol, TCI America™

Manufacturer:  TCI America M09631ML

 View more versions of this product

Catalog No. M09631ML



Specifications

Specifications

(S)-(+)-3-Methyl-1-pentanol
Colorless
42072-39-9
MFCD00135167
IWTBVKIGCDZRPL-LURJTMIESA-N
(3S)-3-methylpentan-1-ol
641003
Clear Liquid at 20°C
152°C
1mL
C6H14O
1987
CCC(C)CCO
102.177
102.18
≥98.0% (GC)

Chemical Identifiers

42072-39-9
102.177
IWTBVKIGCDZRPL-LURJTMIESA-N
(3S)-3-methylpentan-1-ol
C6H14O
MFCD00135167
641003
CCC(C)CCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : No

Documents