missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-(+)-3-Hydroxytetrahydrofuran, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 304430010

 View more versions of this product

Catalog No. AC304430010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

(S)-(+)-3-Hydroxytetrahydrofuran
+18.5° (20°C c=2 MeOH)
1g
98.5
Authentic
1.4490 to 1.4510 (20°C, 589nm)
≥98.5%
+15° to +20° (20°C, 589nm) (c=2, MeOH)
88.11
1.298g/mL
Clear colorless to pale yellow
86087-23-2
100.0
Glass bottle
Liquid
99%
C4H8O2
MFCD00064327

Chemical Identifiers

86087-23-2
100.0
C4H8O2
MFCD00064327
≥98.5%
98.5
99%
88.11
(S)-(+)-3-Hydroxytetrahydrofuran

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.