Please login to your online account to display your discounted pricing

(S)-3-Hydroxyphenylglycine, Tocris

Group I mGlu agonist, active isomer

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material (S)-3-Hydroxyphenylglycine
Assay Percent Range >99%
Molecular Formula C8H9NO3
Formula Weight 167.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

032610

 
r & d systems
RDS032610
10mg Each for $284.81

032650

 
r & d systems
RDS032650
50mg Each for $1,180.32
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 167.16
Chemical Name or Material (S)-3-Hydroxyphenylglycine
Recommended Storage Desiccate at Room Temperature
Assay Percent Range >99%
Molecular Formula C8H9NO3
CAS 71301-82-1
Safety and Handling

Desiccate at Room Temperature