missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(S)-3-Hydroxy-gamma-butyrolactone, TCI America™

$73.33 - $220.00

Chemical Identifiers

CAS 7331-52-4
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00211247
InChI Key FUDDLSHBRSNCBV-VKHMYHEASA-N
Synonym s-3-hydroxy-gamma-butyrolactone, s-4-hydroxydihydrofuran-2 3h-one, 4s-4-hydroxyoxolan-2-one, unii-2lie7314jh, s-4-hydroxytetrahydrofuran-2-one, s-beta-hydroxy-gamma-butyrolactone, 4s-4-hydroxydihydrofuran-2 3h-one, s-4,5-dihydro-4-hydroxy-2 3h-furanone, 2 3h-furanone, dihydro-4-hydroxy-, 4s, 4s-4-hydroxy-3,4,5-trihydrofuran-2-one
PubChem CID 7269389
IUPAC Name (4S)-4-hydroxyoxolan-2-one
SMILES C1C(COC1=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H09395G View Documents TCI America
H09395G
5g Each for $220.00
 
H09391G View Documents TCI America
H09391G
1g Each for $73.33
 
Specifications

Chemical Identifiers

7331-52-4
102.089
FUDDLSHBRSNCBV-VKHMYHEASA-N
7269389
C1C(COC1=O)O
C4H6O3
MFCD00211247
s-3-hydroxy-gamma-butyrolactone, s-4-hydroxydihydrofuran-2 3h-one, 4s-4-hydroxyoxolan-2-one, unii-2lie7314jh, s-4-hydroxytetrahydrofuran-2-one, s-beta-hydroxy-gamma-butyrolactone, 4s-4-hydroxydihydrofuran-2 3h-one, s-4,5-dihydro-4-hydroxy-2 3h-furanone, 2 3h-furanone, dihydro-4-hydroxy-, 4s, 4s-4-hydroxy-3,4,5-trihydrofuran-2-one
(4S)-4-hydroxyoxolan-2-one

Specifications

100°C
5g
C4H6O3
s-3-hydroxy-gamma-butyrolactone, s-4-hydroxydihydrofuran-2 3h-one, 4s-4-hydroxyoxolan-2-one, unii-2lie7314jh, s-4-hydroxytetrahydrofuran-2-one, s-beta-hydroxy-gamma-butyrolactone, 4s-4-hydroxydihydrofuran-2 3h-one, s-4,5-dihydro-4-hydroxy-2 3h-furanone, 2 3h-furanone, dihydro-4-hydroxy-, 4s, 4s-4-hydroxy-3,4,5-trihydrofuran-2-one
C1C(COC1=O)O
102.089
102.09
≥95.0% (GC)
Yellow
7331-52-4
MFCD00211247
FUDDLSHBRSNCBV-VKHMYHEASA-N
(4S)-4-hydroxyoxolan-2-one
7269389
Clear Liquid at 20°C
(S)-3-Hydroxy-gamma-butyrolactone
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6982

TSCA : No